Search
Search

APBB3| Gene Symbol: APBB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006051.3 | NP_006042.3 | 5 | 780 | ||
| NM_133172.2 | NP_573418.2 | 5 | 780 | |||
| NM_133173.2 | NP_573419.2 | 5 | 780 | |||
| NM_133174.2 | NP_573420.2 | 5 | 780 | |||
| GenBank mRNA | AB018247.1 | - | 5 | 472 | ||
| AB049618.1 | - | 5 | 508 | |||
| AB061224.1 | - | 5 | 421 | |||
| AB079687.1 | - | 5 | 421 | |||
| AB094990.1 | - | 5 | 421 | |||
| AB209839.1 | - | 5 | 508 | |||
| AF224708.1 | - | 5 | 441 | |||
| AF224709.1 | - | 5 | 441 | |||
| AF224710.1 | - | 5 | 441 | |||
| AF224711.1 | - | 5 | 441 | |||
| AK074885.1 | - | 2 | 315 | |||
| AK075280.1 | - | 5 | 743 | |||
| AK125244.1 | - | 4 | 1041 | |||
| AK304824.1 | - | 3 | 409 | |||
| BC013158.1 | - | 5 | 780 | 3870148 | ||
| BC125187.1 | - | 5 | 772 | 40122344 | ||