Search
Search

| Gene Symbol: LYPLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007260.2 | NP_009191.1 | 9 | 725 | ||
| XM_005245728.4 | XP_005245785.1 | 9 | 1197 | |||
| XM_017000165.1 | XP_016855654.1 | 9 | 1092 | |||
| GenBank mRNA | AF090423.1 | - | 9 | 771 | ||
| AF098668.1 | - | 9 | 725 | |||
| AK024724.1 | - | 5 | 405 | |||
| BC017034.2 | - | 9 | 690 | 3845764 | ||
| BC017193.2 | - | 9 | 679 | 3138786 | ||
| Gene Symbol: LYPLA2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033250.1 | 1 | 699 | |||