Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5KL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135219.1 | NP_001128691.1 | 10 | 1430 | ||
NM_173492.1 | NP_775763.1 | 5 | 986 | |||
XM_017014279.1 | XP_016869768.1 | 9 | 2478 | |||
GenBank mRNA | BC042184.1 | - | 5 | 986 | 5163193 |