Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGPAT3 SIRNA Assays ›
          Gene Symbol:
          AGPAT3
          Gene Name:
          1-acylglycerol-3-phosphate O-acyltransferase 3
          Gene Aliases:
          LPAAT-GAMMA1, LPAAT3
          Chromosome Location:
          Chr.21: 43865160 - 43987594 on Build GRCh38
          UniGene ID:
          Hs.248785
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s224432
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 562.00
          Your Price
          Online offer: 533.65
          533.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGPAT3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037553.1 NP_001032642.1 8 958
          NM_020132.4 NP_064517.1 9 1078
          XM_005261160.4 XP_005261217.1 9 1079
          XM_006724029.3 XP_006724092.1 8 1007
          XM_006724030.3 XP_006724093.1 8 975
          XM_006724031.3 XP_006724094.1 8 978
          XM_011529660.2 XP_011527962.1 8 817
          XM_011529661.2 XP_011527963.1 8 1132
          XM_011529662.2 XP_011527964.1 10 1782
          XM_011529663.2 XP_011527965.1 8 991
          XM_011529664.2 XP_011527966.1 8 932
          XM_011529665.2 XP_011527967.1 10 1433
          XM_017028408.1 XP_016883897.1 8 961
          XM_017028409.1 XP_016883898.1 8 1039
          XM_017028410.1 XP_016883899.1 8 1044
          XM_017028411.1 XP_016883900.1 8 917
          XM_017028412.1 XP_016883901.1 9 2010
          XM_017028413.1 XP_016883902.1 9 978
          GenBank mRNA AB040138.1 - 8 1033
          AF156774.1 - 9 1039
          AF156775.1 - 8 902
          AK125804.1 - 6 3695
          AL832919.1 - 6 651
          AY358704.1 - 1127
          BC011971.1 - 9 1052 4148915
          BC040603.1 - 10 1782 5270905
          BC063552.1 - 8 1348 4800514

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00220017_m1

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidic acid biosynthetic process
          phospholipid biosynthetic process
          CDP-diacylglycerol biosynthetic process
          triglyceride biosynthetic process
          1-acylglycerol-3-phosphate O-acyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0