Search
Search

SELO| Gene Symbol: SELO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031454.1 | NP_113642.1 | 2 | 659 | ||
| GenBank mRNA | AY324823.1 | - | 2 | 659 | ||
| BC001099.2 | - | 2 | 149 | |||
| BC110866.1 | - | 2 | 629 | 6041782 | ||
| CR456464.1 | - | 2 | 175 | |||