Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD36B SIRNA Assays ›
          Gene Symbol:
          ANKRD36B ANKRD36 ANKRD36C
          Gene Name:
          ankyrin repeat domain 36B
          ankyrin repeat domain 36
          ankyrin repeat domain 36C
          Gene Aliases:
          KIAA1641
          UNQ2430
          -
          Chromosome Location:
          Chr.2: 97492663 - 97590245 on Build GRCh38
          Chromosome Location:
          Chr.2: 97113186 - 97264521 on Build GRCh38
          Chromosome Location:
          Chr.2: 95849548 - 95992320 on Build GRCh38
          UniGene ID:
          Hs.532921
          Hs.541894
          Hs.646318
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s227412

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD36B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_025190.3 NP_079466.3 2 482
          XM_005263995.4 XP_005264052.1 2 762
          XM_006712661.1 XP_006712724.1 2 762
          XM_011511555.1 XP_011509857.1 2 762
          XM_011511556.1 XP_011509858.1 2 762
          XM_011511557.2 XP_011509859.1 2 762
          XM_011511558.1 XP_011509860.1 2 762
          XM_017004578.1 XP_016860067.1 2 762
          XM_017004579.1 XP_016860068.1 2 762
          XM_017004580.1 XP_016860069.1 2 762
          XM_017004581.1 XP_016860070.1 2 762
          XM_017004582.1 XP_016860071.1 2 762
          XM_017004583.1 XP_016860072.1 2 762
          XM_017004584.1 XP_016860073.1 2 762
          XM_017004585.1 XP_016860074.1 2 762
          XM_017004587.1 XP_016860076.1 2 762
          XM_017004588.1 XP_016860077.1 2 762
          XM_017004589.1 XP_016860078.1 2 762
          XM_017004590.1 XP_016860079.1 2 762
          XM_017004592.1 XP_016860081.1 2 762
          XM_017004593.1 XP_016860082.1 2 762
          XM_017004594.1 XP_016860083.1 2 762
          XM_017004595.1 XP_016860084.1 2 762
          GenBank mRNA AK074570.1 - 2 383
          BC071771.1 - 2 308
          BM511319.1 - 2 74
          CR741521.1 - 2 383
          DR001190.1 - 2 371
          Gene Symbol: ANKRD36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164315.1 NP_001157787.1 2 482
          XM_006712514.3 XP_006712577.1 2 792
          XM_011511130.1 XP_011509432.1 2 792
          XM_011511131.1 XP_011509433.1 2 792
          XM_017004009.1 XP_016859498.1 2 792
          XM_017004010.1 XP_016859499.1 2 792
          XM_017004011.1 XP_016859500.1 2 792
          XM_017004012.1 XP_016859501.1 2 792
          XM_017004013.1 XP_016859502.1 2 792
          XM_017004014.1 XP_016859503.1 2 792
          XM_017004015.1 XP_016859504.1 2 792
          XM_017004016.1 XP_016859505.1 2 792
          XM_017004017.1 XP_016859506.1 2 792
          XM_017004018.1 XP_016859507.1 2 792
          XM_017004019.1 XP_016859508.1 2 792
          XM_017004020.1 XP_016859509.1 2 792
          XM_017004021.1 XP_016859510.1 2 792
          XM_017004022.1 XP_016859511.1 2 792
          XM_017004023.1 XP_016859512.1 2 792
          XM_017004024.1 XP_016859513.1 2 792
          XM_017004025.1 XP_016859514.1 2 792
          XM_017004026.1 XP_016859515.1 2 792
          XM_017004027.1 XP_016859516.1 2 792
          XM_017004028.1 XP_016859517.1 2 792
          XM_017004029.1 XP_016859518.1 2 792
          XM_017004030.1 XP_016859519.1 2 792
          XM_017004031.1 XP_016859520.1 2 792
          XM_017004032.1 XP_016859521.1 2 792
          XM_017004033.1 XP_016859522.1 2 792
          XM_017004034.1 XP_016859523.1 2 792
          XM_017004035.1 XP_016859524.1 2 792
          XM_017004036.1 XP_016859525.1 2 792
          XM_017004037.1 XP_016859526.1 2 792
          XM_017004038.1 XP_016859527.1 2 792
          XM_017004039.1 XP_016859528.1 2 792
          XM_017004040.1 XP_016859529.1 2 792
          XM_017004041.1 XP_016859530.1 2 792
          XM_017004042.1 XP_016859531.1 2 792
          XM_017004043.1 XP_016859532.1 2 792
          XM_017004044.1 XP_016859533.1 2 792
          XM_017004045.1 XP_016859534.1 2 792
          XM_017004046.1 XP_016859535.1 2 792
          XM_017004047.1 XP_016859536.1 2 792
          XM_017004048.1 XP_016859537.1 2 792
          XM_017004049.1 XP_016859538.1 2 792
          GenBank mRNA AK304740.1 - 2 422
          AK307629.1 - 2 389
          AK316541.1 - 2 422
          AY358571.1 - 2 481
          BC046186.1 - 2 361
          BC128262.1 - 2 321
          BC128517.1 - 2 321 40118468
          BX647769.1 - 2 427
          Gene Symbol: ANKRD36C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310154.1 NP_001297083.1 2 850
          GenBank mRNA AK302433.1 - 2 324

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding
          ion channel inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0