Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019002.3 | NP_061875.2 | 5 | 1921 | ||
XM_017004376.1 | XP_016859865.1 | 5 | 1914 | |||
XM_017004377.1 | XP_016859866.1 | 5 | 1914 | |||
GenBank mRNA | AJ242682.2 | - | 5 | 1926 | ||
AK025155.1 | - | 4 | 1454 | |||
AK026300.1 | - | 1 | 367 | |||
BC033075.1 | - | 5 | 1893 | |||
BC040001.1 | - | 5 | 1853 | 5556133 |