Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPP8 SIRNA Assays ›
Gene Symbol:
DPP8
Gene Name:
dipeptidyl peptidase 8
Gene Aliases:
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
Chromosome Location:
Chr.15: 65442463 - 65517713 on Build GRCh38
UniGene ID:
Hs.458609
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29656
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPP8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320875.1
NP_001307804.1
19
2687
NM_001320876.1
NP_001307805.1
18
2656
NM_017743.5
NP_060213.2
16
2251
NM_130434.4
NP_569118.1
18
2551
NM_197960.3
NP_932064.1
19
2592
NM_197961.3
NP_932065.1
18
2534
NR_135485.1
19
2739
NR_135486.1
20
2722
XM_005254505.1
XP_005254562.1
10
1233
XM_011521730.1
XP_011520032.1
17
3811
XM_011521731.1
XP_011520033.1
17
3766
XM_011521732.2
XP_011520034.1
18
3940
XM_011521733.1
XP_011520035.1
17
2358
XM_011521734.1
XP_011520036.1
16
3640
XM_017022373.1
XP_016877862.1
18
3940
XM_017022374.1
XP_016877863.1
17
3787
XM_017022375.1
XP_016877864.1
17
2403
XM_017022376.1
XP_016877865.1
17
2379
XM_017022377.1
XP_016877866.1
16
2229
XM_017022378.1
XP_016877867.1
17
3787
XM_017022379.1
XP_016877868.1
16
2205
XM_017022380.1
XP_016877869.1
17
2358
XM_017022381.1
XP_016877870.1
15
2076
XM_017022382.1
XP_016877871.1
11
1360
XM_017022383.1
XP_016877872.1
11
1332
GenBank mRNA
AF175225.1
-
6
686
AF176779.1
-
6
683
AF221634.1
-
18
2525
AF221635.1
-
5
598
AF221636.1
-
9
1062
AF221637.1
-
9
1065
AK000290.1
-
16
2173
AK027826.1
-
13
1577
AK291057.1
-
19
2662
AK296915.1
-
18
2627
AK302638.1
-
20
2658
AY172659.1
-
17
2319
AY354202.1
-
19
2520
BC030688.2
-
18
2525
4822550
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028906, AK016546, BC043124, BC059222, AK155605, and others.
Matching Taqman Gene Expression Assay:
Hs00214745_m1
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
immune response
serine-type peptidase activity
dipeptidyl-peptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-89mxr:80/10.239.163.131:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0