Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NDRG2 SIRNA Assays ›
          Gene Symbol:
          NDRG2
          Gene Name:
          NDRG family member 2
          Gene Aliases:
          SYLD
          Chromosome Location:
          Chr.14: 21016763 - 21070872 on Build GRCh38
          UniGene ID:
          Hs.525205
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s33035

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NDRG2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282211.1 NP_001269140.1 7 496
          NM_001282212.1 NP_001269141.1 8 770
          NM_001282213.1 NP_001269142.1 8 775
          NM_001282214.1 NP_001269143.1 8 763
          NM_001282215.1 NP_001269144.1 8 593
          NM_001282216.1 NP_001269145.1 7 511
          NM_001320329.1 NP_001307258.1 8 1428
          NM_016250.2 NP_057334.1 8 618
          NM_201535.1 NP_963293.1 9 660
          NM_201536.1 NP_963294.1 7 551
          NM_201537.1 NP_963831.1 8 593
          NM_201538.1 NP_963832.1 7 528
          NM_201539.1 NP_963833.1 8 570
          NM_201540.1 NP_963834.1 9 637
          NM_201541.1 NP_963835.1 8 595
          XM_011536996.2 XP_011535298.1 9 647
          XM_011536997.1 XP_011535299.1 9 648
          XM_011536998.1 XP_011535300.1 9 638
          XM_011536999.1 XP_011535301.1 7 1043
          XM_011537001.1 XP_011535303.1 8 601
          XM_011537002.1 XP_011535304.1 8 747
          XM_017021480.1 XP_016876969.1 9 599
          XM_017021481.1 XP_016876970.1 9 638
          XM_017021482.1 XP_016876971.1 9 647
          XM_017021483.1 XP_016876972.1 8 531
          XM_017021484.1 XP_016876973.1 8 573
          XM_017021485.1 XP_016876974.1 9 638
          XM_017021486.1 XP_016876975.1 9 648
          XM_017021487.1 XP_016876976.1 8 557
          XM_017021488.1 XP_016876977.1 8 598
          XM_017021489.1 XP_016876978.1 8 703
          XM_017021490.1 XP_016876979.1 7 495
          XM_017021491.1 XP_016876980.1 7 530
          XM_017021492.1 XP_016876981.1 8 601
          XM_017021493.1 XP_016876982.1 8 747
          XM_017021494.1 XP_016876983.1 9 599
          XM_017021495.1 XP_016876984.1 8 531
          XM_017021496.1 XP_016876985.1 7 530
          XM_017021497.1 XP_016876986.1 3 128
          GenBank mRNA AB033074.1 - 7 480
          AF087872.1 - 7 534
          AF159092.3 - 8 541
          AF304051.1 - 9 660
          AK057843.1 - 8 564
          AK057847.1 - 7 528
          AK090657.1 - 7 467
          AK096999.1 - 8 1428
          AK130029.1 - 7 466
          AK293514.1 - 7 496
          AL136574.1 - 7 505
          BC010458.1 - 7 499 4215141
          BC011240.1 - 8 553 4153602
          BC013209.1 - 1 106
          BC093038.1 - 8 594 4811718
          BG818977.1 - 8 574
          BI759006.1 - 9 579
          BM810623.1 - 8 534
          BX247987.1 - 8 629
          BX647091.1 - 5 693
          BX647748.1 - 8 710
          CB153966.1 - 7 594
          CR749252.1 - 8 763

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cytokine production
          signal transduction
          regulation of vascular endothelial growth factor production
          Wnt signaling pathway
          substantia nigra development
          cell differentiation
          negative regulation of smooth muscle cell proliferation
          negative regulation of ERK1 and ERK2 cascade
          regulation of platelet-derived growth factor production
          molecular_function
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0