Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302643.1 | NP_001289572.1 | 6 | 840 | ||
NM_022082.3 | NP_071365.3 | 5 | 778 | |||
XM_011528978.2 | XP_011527280.1 | 5 | 690 | |||
GenBank mRNA | AK074107.1 | - | 5 | 714 | ||
AK095473.1 | - | 5 | 629 | |||
AK125597.1 | - | 6 | 783 | |||
AK298503.1 | - | 5 | 684 | |||
BC025312.1 | - | 6 | 780 | 4563089 | ||
BC038593.1 | - | 5 | 620 |