Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLG4 SIRNA Assays ›
          Gene Symbol:
          DLG4
          Gene Name:
          discs large MAGUK scaffold protein 4
          Gene Aliases:
          PSD95, SAP-90, SAP90
          Chromosome Location:
          Chr.17: 7189890 - 7220050 on Build GRCh38
          UniGene ID:
          Hs.463928
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4130
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DLG4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128827.2 NP_001122299.1 6 828
          NM_001321074.1 NP_001308003.1 8 1676
          NM_001321075.1 NP_001308004.1 6 837
          NM_001321076.1 NP_001308005.1 5 482
          NM_001321077.1 NP_001308006.1 5 473
          NM_001365.4 NP_001356.1 8 1685
          NR_135527.1 8 1685
          XM_005256491.1 XP_005256548.1 6 927
          XM_011523698.1 XP_011522000.1 8 1573
          XM_011523699.2 XP_011522001.1 9 815
          XM_011523702.1 XP_011522004.1 5 352
          XM_017024288.1 XP_016879777.1 5 573
          XM_017024289.1 XP_016879778.1 5 564
          XM_017024290.1 XP_016879779.1 5 1042
          GenBank mRNA AF028825.1 - 6 387
          AK293835.1 - 6 834
          AK296357.1 - 8 1373
          AK297346.1 - 5 430
          AK299801.1 - 5 444
          AK307804.1 - 6 811
          AK308533.1 - 5 587
          AK316096.1 - 6 390
          BC136553.1 - 8 1209 9020543
          BP213421.1 - 5 473
          U68138.1 - 4 244
          U83192.1 - 8 1343

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019621, XM_006246686, XM_006246689, XM_006246681, XM_006246687, and others.

          Matching Taqman Gene Expression Assay:


          Hs00176354_m1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          negative regulation of receptor internalization
          protein complex assembly
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          chemical synaptic transmission
          nervous system development
          learning
          synaptic vesicle maturation
          ion transmembrane transport
          social behavior
          protein localization to synapse
          locomotory exploration behavior
          positive regulation of GTPase activity
          establishment of protein localization
          establishment or maintenance of epithelial cell apical/basal polarity
          GMP metabolic process
          GDP metabolic process
          regulation of long-term neuronal synaptic plasticity
          positive regulation of synaptic transmission
          neuromuscular process controlling balance
          dendritic spine morphogenesis
          vocalization behavior
          AMPA glutamate receptor clustering
          receptor localization to synapse
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          regulation of grooming behavior
          guanylate kinase activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          protein C-terminus binding
          ligand-gated ion channel activity
          kinase binding
          protein phosphatase binding
          PDZ domain binding
          beta-1 adrenergic receptor binding
          D1 dopamine receptor binding
          P2Y1 nucleotide receptor binding
          protein complex binding
          acetylcholine receptor binding
          ionotropic glutamate receptor binding
          neuroligin family protein binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0