Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193476.1 | NP_001180405.1 | 10 | 1366 | ||
NM_052961.3 | NP_443193.1 | 10 | 1390 | |||
NM_138718.2 | NP_619732.2 | 8 | 1113 | |||
XM_011514294.2 | XP_011512596.1 | 10 | 1609 | |||
XM_017010235.1 | XP_016865724.1 | 10 | 1416 | |||
GenBank mRNA | AF314959.1 | - | 9 | 1260 | ||
AF331522.1 | - | 10 | 1334 | |||
AK097410.1 | - | 4 | 452 | |||
AK122928.1 | - | 10 | 1366 | |||
AK125189.1 | - | 10 | 1610 | |||
BC025408.2 | - | 8 | 1106 | 4823098 |