Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DRD3 SIRNA Assays ›
          Gene Symbol:
          DRD3
          Gene Name:
          dopamine receptor D3
          Gene Aliases:
          D3DR, ETM1, FET1
          Chromosome Location:
          Chr.3: 114127623 - 114199407 on Build GRCh38
          UniGene ID:
          Hs.121478
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4289

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DRD3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000796.5 NP_000787.2 5 1084
          NM_001282563.2 NP_001269492.1 6 943
          NM_001290809.1 NP_001277738.1 6 1076
          NM_033663.5 NP_387512.3 5 1084
          XM_017005829.1 XP_016861318.1 6 904
          GenBank mRNA AK292309.1 - 6 943
          BC095510.1 - 4 654 7001986
          BC128122.1 - 6 998 40112299
          BC128123.1 - 5 730 40112300
          KJ534829.1 - 4 653
          KJ535032.1 - 3 540
          L20469.1 - 4 653
          U32499.1 - 4 653

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          synaptic transmission, dopaminergic
          response to amphetamine
          regulation of blood volume by renin-angiotensin
          G-protein coupled receptor internalization
          cellular calcium ion homeostasis
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          negative regulation of adenylate cyclase activity
          adenylate cyclase-inhibiting dopamine receptor signaling pathway
          learning or memory
          learning
          locomotory behavior
          positive regulation of cell proliferation
          visual learning
          regulation of dopamine secretion
          regulation of lipid metabolic process
          regulation of cAMP metabolic process
          negative regulation of sodium:proton antiporter activity
          positive regulation of cytokinesis
          circadian regulation of gene expression
          response to histamine
          social behavior
          gastric emptying
          positive regulation of renal sodium excretion
          regulation of multicellular organism growth
          response to cocaine
          dopamine metabolic process
          response to drug
          response to morphine
          regulation of circadian sleep/wake cycle, sleep
          response to ethanol
          negative regulation of blood pressure
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          acid secretion
          behavioral response to cocaine
          negative regulation of oligodendrocyte differentiation
          arachidonic acid secretion
          negative regulation of protein secretion
          musculoskeletal movement, spinal reflex action
          regulation of dopamine uptake involved in synaptic transmission
          negative regulation of protein kinase B signaling
          prepulse inhibition
          negative regulation of dopamine receptor signaling pathway
          positive regulation of dopamine receptor signaling pathway
          regulation of locomotion involved in locomotory behavior
          dopamine neurotransmitter receptor activity, coupled via Gs
          dopamine neurotransmitter receptor activity, coupled via Gi/Go
          protein binding
          drug binding
          protein domain specific binding
          D1 dopamine receptor binding
          dopamine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pslnx:80/100.66.77.47:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0