Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELN SIRNA Assays ›
Gene Symbol:
ELN
Gene Name:
elastin
Gene Aliases:
SVAS, WBS, WS
Chromosome Location:
Chr.7: 74027772 - 74069907 on Build GRCh38
UniGene ID:
Hs.647061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s531158
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000501.3
NP_000492.2
33
2630
NM_001081752.2
NP_001075221.1
30
2489
NM_001081753.2
NP_001075222.1
31
2534
NM_001081754.2
NP_001075223.1
32
2591
NM_001081755.2
NP_001075224.1
32
2573
NM_001278912.1
NP_001265841.1
32
2576
NM_001278913.1
NP_001265842.1
29
2387
NM_001278914.1
NP_001265843.1
31
2558
NM_001278915.1
NP_001265844.1
33
2648
NM_001278916.1
NP_001265845.1
29
2432
NM_001278917.1
NP_001265846.1
32
2600
NM_001278918.1
NP_001265847.1
28
2309
NM_001278939.1
NP_001265868.1
34
2816
XM_005250187.1
XP_005250244.1
32
2271
XM_005250188.1
XP_005250245.1
32
2263
XM_011515868.1
XP_011514170.1
33
2331
XM_011515869.1
XP_011514171.1
32
2260
XM_011515870.1
XP_011514172.1
32
2626
XM_011515871.1
XP_011514173.1
32
2289
XM_011515872.1
XP_011514174.1
32
2277
XM_011515873.1
XP_011514175.1
32
2274
XM_011515874.1
XP_011514176.1
31
2596
XM_011515875.1
XP_011514177.1
31
2252
XM_011515876.1
XP_011514178.1
31
2232
XM_011515877.1
XP_011514179.1
31
2220
XM_017011813.1
XP_016867302.1
31
2174
XM_017011814.1
XP_016867303.1
31
2232
GenBank mRNA
AB208942.1
-
32
2226
AK075554.1
-
29
2070
AK092232.1
-
28
1924
AK094138.1
-
33
2063
AK095990.1
-
31
2154
AK122731.1
-
25
1782
AK125511.1
-
31
2216
AK130894.1
-
18
1125
AK225659.1
-
32
2187
BC035570.1
-
33
2235
BC050379.1
-
26
1504
BC065566.1
-
29
2063
6153564
BX537939.1
-
32
2211
BX538199.1
-
30
2121
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: BC085910, J04035.
This siRNA also targets the following transcripts in Mouse: BC034057, BC051074, AK041860, AK080277, BC051649, and others.
Panther Classification:
Molecular Function -
structural protein
Gene Ontology Categories:
Function(s)
Process(es)
respiratory gaseous exchange
blood circulation
cell proliferation
organ morphogenesis
extracellular matrix disassembly
extracellular matrix organization
extracellular matrix structural constituent
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0