Search
Search

Bmp10| Gene Symbol: Bmp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009756.2 | NP_033886.2 | 2 | 1224 | ||
| GenBank mRNA | AF101033.1 | - | 2 | 1190 | ||
| BC145044.1 | - | 2 | 1224 | 9053592 | ||
| BC145983.1 | - | 2 | 1224 | 40131286 | ||