Search
Search

Galnt4| Gene Symbol: Galnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015737.4 | NP_056552.1 | 1 | 1462 | ||
| GenBank mRNA | AK033494.1 | - | 1 | 1427 | ||
| AK148036.1 | - | 1 | 1430 | |||
| AK153253.1 | - | 1 | 1440 | |||
| BC057882.1 | - | 1 | 1453 | 5350623 | ||
| U73819.1 | - | 1 | 1182 | |||