Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013555.4 | NP_038583.1 | 1 | 1062 | ||
GenBank mRNA | AK134427.1 | - | 1 | 852 | ||
AK144746.1 | - | 1 | 207 | |||
BC019150.1 | - | 1 | 1097 | 5064578 |