Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS3 SIRNA Assays ›
          Gene Symbol:
          Rps3
          Gene Name:
          ribosomal protein S3
          Gene Aliases:
          D7Ertd795e, Rs_3
          Chromosome Location:
          Chr.7: 99477896 - 99483709 on Build GRCm38
          UniGene ID:
          Mm.236868
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s77500

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012052.2 NP_036182.1 3, 4 344
          GenBank mRNA AK003181.1 - 3, 4 282
          AK010579.1 - 3, 4 287
          AK010678.1 - 3, 4 285
          AK011655.1 - 3, 4 282
          AK012234.1 - 3, 4 286
          AK012314.1 - 3, 4 286
          AK012538.1 - 3, 4 286
          AK051243.1 - 3, 4 287
          AK088038.1 - 3, 4 287
          AK143603.1 - 3, 4 286
          AK146165.1 - 3, 4 287
          AK146796.1 - 3, 4 287
          AK148030.1 - 3, 4 286
          AK150479.1 - 3, 4 280
          AK159508.1 - 3, 4 287
          AK160526.1 - 3, 4 286
          AK164077.1 - 3, 4 287
          AK165174.1 - 3, 4 276
          AK166750.1 - 3, 4 288
          AK167210.1 - 3, 4 286
          AK167266.1 - 3, 4 276
          AK168845.1 - 3, 4 276
          AK206775.1 - 2, 3 241
          BC010721.1 - 3, 4 274 2811930
          CJ074413.1 - 3, 4 374
          U89414.1 - 1, 2 62
          X76772.1 - 3, 4 261

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA metabolic process
          DNA repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          translation
          regulation of translation
          apoptotic process
          cellular response to DNA damage stimulus
          cell cycle
          mitotic nuclear division
          negative regulation of DNA repair
          positive regulation of NF-kappaB transcription factor activity
          cell division
          nucleic acid phosphodiester bond hydrolysis
          positive regulation of DNA N-glycosylase activity
          positive regulation of apoptotic signaling pathway
          nucleic acid binding
          DNA binding
          damaged DNA binding
          RNA binding
          mRNA binding
          structural constituent of ribosome
          DNA-(apurinic or apyrimidinic site) lyase activity
          protein binding
          oxidized purine nucleobase lesion DNA N-glycosylase activity
          lyase activity
          enzyme binding
          protein kinase binding
          poly(A) RNA binding
          protein kinase A binding
          NF-kappaB binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0