Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170591.1 | NP_001164062.1 | 6 | 598 | ||
NM_020045.3 | NP_064429.2 | 6 | 595 | |||
GenBank mRNA | AJ132616.1 | - | 6 | 526 | ||
AK002244.1 | - | 6 | 544 | |||
AK003730.1 | - | 6 | 543 | |||
AK145558.1 | - | 6 | 599 | |||
AY335195.1 | - | 6 | 502 | |||
BC018355.1 | - | 6 | 516 | 3584656 |