Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134093.2 | NP_598854.1 | 2 | 229 | ||
GenBank mRNA | AF287293.1 | - | 2 | 169 | ||
AF401483.1 | - | 2 | 62 | |||
AK032864.1 | - | 1 | 586 | |||
AK041312.1 | - | 1 | 558 | |||
AK045256.1 | - | 2 | 49 | |||
AK046496.1 | - | 2 | 149 | |||
AK080221.1 | - | 2 | 149 | |||
AK154676.1 | - | 2 | 144 | |||
AK158718.1 | - | 2 | 122 | |||
AK169341.1 | - | 2 | 149 | |||
BY293457.1 | - | 2 | 229 |