Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl9l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030256.2 | NP_084532.2 | 7 | 2327 | ||
GenBank mRNA | AB128033.1 | - | 6 | 2113 | ||
AK155681.1 | - | 8 | 2287 | |||
AY296058.1 | - | 7 | 2327 | |||
BC082304.1 | - | 6 | 2461 | 6856460 |