Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTN1 SIRNA Assays ›
          Gene Symbol:
          ACTN1
          Gene Name:
          actinin alpha 1
          Gene Aliases:
          BDPLT15
          Chromosome Location:
          Chr.14: 68874123 - 68979366 on Build GRCh38
          UniGene ID:
          Hs.235750
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s967

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACTN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001102.3 NP_001093.1 6 878
          NM_001130004.1 NP_001123476.1 6 878
          NM_001130005.1 NP_001123477.1 6 878
          XM_011537265.2 XP_011535567.1 6 670
          XM_011537266.2 XP_011535568.1 6 670
          XM_011537267.2 XP_011535569.1 6 670
          XM_011537268.2 XP_011535570.1 6 670
          XM_017021720.1 XP_016877209.1 6 670
          XM_017021721.1 XP_016877210.1 6 670
          XM_017021722.1 XP_016877211.1 6 815
          XM_017021723.1 XP_016877212.1 6 815
          XM_017021725.1 XP_016877214.1 6 989
          XM_017021726.1 XP_016877215.1 6 989
          XM_017021727.1 XP_016877216.1 6 815
          XM_017021728.1 XP_016877217.1 6 989
          GenBank mRNA AK225782.1 - 6 775
          AK295099.1 - 6 774
          AK298496.1 - 6 653
          AK299362.1 - 6 516
          BC003576.1 - 6 810 3547017
          BC015766.1 - 6 685 4856734
          BT007207.1 - 6 568
          BX248020.1 - 6 541
          DQ496098.1 - 6 751
          EU716325.1 - 6 588
          FJ410030.1 - 6 588
          M95178.1 - 6 679
          X15804.1 - 6 766

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          actin filament organization
          platelet activation
          platelet formation
          platelet morphogenesis
          regulation of apoptotic process
          focal adhesion assembly
          actin filament bundle assembly
          negative regulation of cellular component movement
          actin filament network formation
          actin crosslink formation
          platelet aggregation
          regulation of nucleic acid-templated transcription
          double-stranded RNA binding
          integrin binding
          calcium ion binding
          protein binding
          vinculin binding
          ligand-dependent nuclear receptor transcription coactivator activity
          protein homodimerization activity
          ion channel binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0