ThermoFisher Scientific
Search
clear search
Recent searches Clear History
Suggestions
All Results
clear search
Search
Search Suggestions
Recent searches Clear History
お問い合わせ
ImmunoCAP ECP Test
Marker of Airway Inflammation
本ページで紹介している製品は、日本では薬機法上未承認であり、国内での販売・提供は行っておりません。
最新の製品情報はグローバルサイトをご参照ください。
The products described on this page are not approved under the Japanese regulatory framework and are not available for sale or distribution in Japan.
Please refer to the global website for the latest product information.
ImmunoCAP™ ECP measures the level of Eosinophil Cationic Protein (ECP) in serum. Eosinophils are the cells chiefly responsible for producing the inflammation associated with some types of asthma. When eosinophils in the airway are activated, they undergo degranulation and release ECP, causing airway epithelial damage. The level of ECP correlates with the level of inflammation in asthmatic patients.
Asthmatic patients with eosinophilic inflammation have elevated levels of ECP.1,2
Regular measurement of ECP may help keep asthma under control
ImmunoCAP ECP can help you identify the severity of the airway inflammation1 and aids in determining appropriate therapy for patients with asthma.
Accurate diagnosis
  • Rule in or rule out if eosinophilic inflammation is involved in the disease and if the patient is responsive to inhaled corticosteroid (ICS) therapy.1,2
Assessment of risk
  • Determine severity of asthma and airway inflammation.1-3
  • Identify the potential risk for having asthma exacerbations.2,4
Relevant treatment decisions
  • A guide in targeting the correct dosage of ICS therapy. 2,5,6
  • Follow-up on patient compliance in anti-inflammatory asthma therapy.2
ImmunoCAP ECP tests
View Catalog
Article Number 14-4515-01
Product Type テスト
Barcode C73
Size 16
Product Info
Article Number 14-4515-02
Barcode C70
Size 16
Product Info
Article Number 14-4515-41
Product Type テスト
Barcode DR3
Size 16
Product Info
Article Number 10-9321-01
Product Type Reagent
Barcode A3Z
Size 2 x 50
Product Info
Article Number 10-9321-02
Product Type Reagent
Barcode C71
Size 2 x 50
Product Info
Article Number 10-9321-41
Product Type Reagent
Barcode CG4
Size 2 x 50
Product Info
References
  1. Directions for use. ImmunoCAP ECP.
  2. Koh, G.C., et al., Eosinophil cationic protein: is it useful in asthma? A systematic review. Respir Med, 2007. 101(4): p. 696-705.
  3. Parra, A., et al., Serum ECP levels in asthmatic patients: comparison with other follow-up parameters. Allergy Asthma Proc, 1996. 17(4): p. 191-7.
  4. Belda, J., et al., Mild exacerbations and eosinophilic inflammation in patients with stable, well-controlled asthma after 1 year of follow-up. Chest, 2001. 119(4): p. 1011-7.
  5. Kristjansson, S., et al., Urinary eosinophil protein X in children with atopic asthma: a useful marker of antiinflammatory treatment. J Allergy Clin Immunol, 1996. 97(6): p. 1179-87.
  6. Lonnkvist, K., et al., Eosinophil markers in blood, serum, and urine for monitoring the clinical course in childhood asthma: impact of budesonide treatment and withdrawal. J Allergy Clin Immunol, 2001. 107(5): p. 812-7.