Search
Simply click the general LC symptom that you are observing, then navigate the symptom details to define potential causes of your problems and receive appropriate solutions to resolve the problem.
Download our LC troubleshooting mobile app for more troubleshooting topic ›
Flat-topped peaks | |
|---|---|
| Detector overload. | Reduce the sample concentration. |
| Detector set-up. | Check the detector attenuation and re-zero. |