Transmission Electron Microscopy Techniques for the Life Sciences
TEM techniques for the life sciences, biomedical research, and drug development
Transmission electron microscopes (TEMs) support a wide variety of applications including cellular and subcellular structural analysis, characterization of pathology and disease mechanisms, drug development and pharmacology, molecular and structural biology, and toxicology studies.
Thermo Scientific TEMs are equipped with the techniques you need for life science research, biomedical analysis, and pharmaceutical drug discovery and development.
2D ultrastructural imaging of cells and tissues
2D imaging with transmission electron microscopy provides some of the highest resolution imaging possible for resin-embedded or cryogenically prepared thin sections of cells and tissues, revealing intracellular structures at exceptional resolution. Thermo Scientific TEMs can also be used in STEM (scanning transmission electron microscope) mode to observe larger areas or further increase image contrast.
Ultrastructural imaging shows the detailed organization of organelles, membranes, and other intracellular components. It is a common technique in pathology and biomedical studies, where it is used to identify and characterize abnormalities in tissue samples. TEM is used to study the development and differentiation of tissues, providing insights into the processes of growth and development. In studies involving nanoparticles, TEM can also be used to observe the interaction of these particles with biological tissues, assessing their distribution, localization, and potential toxic effects.
TEM imaging of muscle tissue. The left image has a pixel size of 14.7 nm. The center image is a higher magnification area of the cyan region of interest (ROI) from the left image, with a pixel size of 4.1 nm. The right image is an even higher magnification of the orange ROI, with a pixel size of 0.82 nm.
Electron tomography at nanometer resolution
TEM electron tomography delivers detailed 3D structural information for a sample at the nanometer scale. This technique involves taking a series of 2D images at different tilt angles and then reconstructing them into a 3D model using computational algorithms.
It can provide detailed structural analyses of cellular structures, including organelles, the cytoskeleton, and membranes, as well as organelle morphology. Electron tomography is also used to visualize the spatial arrangement of proteins and large macromolecular complexes.
3D reconstruction of lung cilia. Data acquired using TEM tomography, with segmentation performed in Thermo Scientific Amira Software.
Cryo-electron tomography
When this technique is combined with a TEM capable of analyzing cryogenic samples, it is possible to leverage tomography for the visualization of biological samples at near-native (vitrified) conditions. This technique, termed cryo-electron tomography, or cryo-ET, is offered on some models of the Thermo Scientific specialized cryo-TEMs, which are designed for cryo-electron microscopy applications.
Correlative light and electron microscopy
Combining the high-resolution structural information of electron microscopy with the contextual information of fluorescence light microscopy, correlative light and electron microscopy (CLEM) is a versatile and powerful technique in the life sciences that bridges the gap between the capabilities of both methods. This integrative approach provides a more comprehensive understanding of biological processes and structures.
CLEM with Thermo Scientific TEMs is powered by easy-to-use software, making data collection and correlation both accurate and efficient.
Multi-scale TEM imaging of fluorescently-labeled cells from a correlative light and electron microscopy workflow.
EDS elemental analysis
Elemental analysis with EDS (energy-dispersive X-ray spectroscopy) is a technique that can be performed in an electron microscope through the addition of a specialized EDS detector, and is widely used to determine the elemental composition of materials.
For life science researchers, EDS can be leverage to identify the presence of specific elements in cells and tissues, or for mineralization studies of bones and teeth. Pharmaceutical researchers use EDS analysis to investigate the elemental composition of drug formulations, or as a tool for contaminant detection. EDS is widely used in materials science, chemistry, and geology research, making this technique a useful addition to transmission electron microscopes that are shared between multiple departments.
Compositional analysis of Magnetospirillum gryphiswaldense, unstained dehydrated cells on TEM grid. Sample courtesy of Prof. Dr. Dirk Schüler, University of Bayreuth, Germany.
Negative stain imaging
Negative stain imaging is a technique used to enhance the contrast of biological specimens such as viruses, proteins, and other macromolecules, making them more visible under the TEM. The electron dense stain surrounds the sample, creating a high-contrast envelope, or outline, against the background. This rapid and relatively simple method of contrast enhancement allows for the visualization of shape and surface details for a sample, even at relatively low electron doses, which are often needed  to preserve the integrity of delicate biological structures.
Negative stain imaging with TEM can be useful as a standalone technique or to assess the quality of protein samples for subsequent cryo-EM single particle analysis.
Negatively stained bacteriophage with a long tail, imaged with transmission electron microscopy.
Nanoparticle characterization for pharmaceutical research
The characterization of nanoparticles by negative-stain TEM or cryo-TEM provides detailed insights into their structure, size, morphology, and surface properties. These attributes are critical for pharmaceutical researchers who are trying to accurately assess attributes such as particle size distribution, morphology, empty/full classification, or particle aggregation.
Thermo Scientific TEM workflows ensure streamlined and accurate data collection and analysis for reduced time to results.
TEM analysis of empty and full AAV2 vectors, used to ensure the quality and efficacy of AAV2-based gene-therapy products.
Cryo-electron microscopy
Cryo-electron microscopy (cryo-EM) is a collection of techniques that enables direct visualization of biological macromolecules, and the observation of biological systems, in their near-native states without the risk of artifacts from chemical fixation or negative staining. It encompasses single particle analysis for the structural analysis of proteins and complexes, cryo-electron tomography for cellular/subcellular imaging, and microcrystal electron diffraction for the characterization of small molecules and compounds. Cryo-EM techniques can access a broad range of biological samples, including large protein complexes, molecular machines, and virus particles, which have traditionally been challenging for other analytical techniques.
Cryo-EM is available as an additional option for all Thermo Scientific TEMs, or as standard in our line of specialized cryo-TEMs, designed to specifically expand your cryo-EM capabilities.
AI-powered segmentation with Amira Software reveals fine details of proteoliposome architecture. Sample courtesy of Dr. Sara García-Linares and Dr. Alvaro Martínez-del-Pozo, Complutense University of Madrid, Spain, and Dr. Jaime Martín-Benito Romero, CNB-CSIC, Spain.
What transmission electron microscope is right for you?
Thermo Scientific TEMs feature enhanced imaging and analysis through simplified and automated operation, improving data quality and acquisition times. They are suited for multi-user and multi-discipline environments, often combining a myriad of applications in a single system.
For Research Use Only. Not for use in diagnostic procedures.