SECURE-Spin: Advanced User Management and Centralized Monitoring Features on Thermo Scientific General Purpose Pro Centrifuges
SECURE-Spin: Advanced User Management and Centralized Monitoring Features on Thermo Scientific General Purpose Pro Centrifuges
SECURE-Spin adds advanced features to the centrifuge, including but not limited to: Secure Mode involving user rights and data management, connection to Centri-Log Plus1 data management software (Cat. No. 75007742) via network, and GMP Mode guiding user through a GMP workflow.
Have Questions?
Catalog NumberMax. RCF
7500999025,830 x g
Catalog number 75009990
Price (USD)
-
Max. RCF:
25,830 x g
In regulated environments, it is crucial to assign user rights and maintain data management in a 21 CFR Part 11-compliant manner. This ensures traceable documentation in order to maintain sample and process safety, to provide audit trails and to record data in a secure way. SECURE-Spin takes the Thermo Scientific General Purpose Pro Centrifuge to the next level with accurate data collection, reporting and archiving as well as management and protection of process data.
In regulated environments, it is crucial to assign user rights and maintain data management in a 21 CFR Part 11-compliant manner. This ensures traceable documentation in order to maintain sample and process safety, to provide audit trails and to record data in a secure way. SECURE-Spin takes the  Thermo Scientific General Purpose Pro Centrifuge to the next level with accurate data collection, reporting and archiving as well as management and protection of process data. 
Specifications
VentilatedYes
CapacityFor both 4 L capacity and 1.6 L capacity
Max. RCF25,830 x g
Max. Speed15,200 rpm
Product LineSorvall
RefrigeratedYes
Unit SizeEach