Spinnaker™ 3 Microplate Mover
Spinnaker™ 3 Microplate Mover
Spinnaker™ 3 Microplate Mover
Spinnaker™ 3 Microplate Mover
Spinnaker™ 3 Microplate Mover
Thermo Scientific™
Spinnaker™ 3 Microplate Mover
Increase lab productivity through add-and-go automation of scientific workflows with the Thermo Scientific™ Spinnaker™ 3 Microplate Mover, featuring patented auto-leveling capabilities.
Have Questions?
Catalog NumberHeight (Metric)
F02360800 mm
Catalog number F02360
Price (USD)
-
Height (Metric):
800 mm
Automate your scientific workflow easily with the Thermo Scientific™ Spinnaker™ 3 Microplate Mover robot. This latest addition to our add-and-go lab automation ecosystem promotes automation adoption by users of any experience level through unmatched ease of use and reliability.
With its innovative auto-leveling gripper and vision based position teaching, the Spinnaker 3 microplate mover offers self-correcting precision in our non-flat world. This designed-for-lab plate mover is well suited for a wide variety of research fields including synthetic biology, drug discovery, and biotechnology.
Achieve higher returns from your lab with reduced effort and investments by adding this latest lab productivity tool. Designed specifically for life science research, the Spinnaker 3 microplate mover robot provides an extremely user-friendly experience. Equipped with the newest auto-leveling capabilities, this version of our long-trusted Spinnaker mover enables adaptation to the non-flat world while lowering user effort. Spinnaker 3 makes automation easier while maintaining a collaborative design, promoting confidence for users working alongside automation and increasing utilization of lab resources, resulting in increased productivity.
Features
  • Auto-leveling gripper
  • Machine vision-based consistent teaching
  • Touchless re-teaching/corrections
  • Automated re-teaching for moved SmartCarts and instruments
  • On-the-fly barcode reads
Reliable
  • Error-proof Smart Cart Docking
  • Collision detection and recovery
  • Automated plate detection
  • Position retention on power failure
Flexible Plate Handling
  • Handles most SBS format microplates
  • Deep well plate
  • Tip boxes
  • SBS tube racks
  • Random or sequential access storage
  • Off-set grip standard
  • Overhead grip optional
Easy-to-use and Teach
  • Step-by-step guided vision-based teaching
  • SmartMove™ technology avoids obstacles
Software
  • Operates with Momentum and other scheduling software
Safety
  • Accordance with Category 3 safety standards
  • Ability to limp without power supply
Accessories
  • Arrayed plate storage
  • Radial plate storage
  • Carousels (5 and 8 column available)
  • BenchTrak™
Specifications
DescriptionReach: 597 mm
Height (Metric)800 mm
Humidity20 to 80% Non-condensing
Nominal Payload (Metric)250 g, 320 g max.
Repeatability±0.25 mm
Temperature (Metric) OperatingNominal: 19 to 25 °C (4 to 40 °C)
Unit SizeEach
Frequently asked questions (FAQs)
Are the Multidrop Combi reagent dispensers compatible with Momentum Workflow Scheduling Software?
Yes, the Multidrop Combi reagent dispensers are compatible with the Momentum Software and can be integrated into automated workflow platforms. If you are planning to set up an automated workflow using the Momentum Software, you will also need a microplate mover such as the Orbitor RS2 Microplate Mover, Spinnaker Microplate Robot, or Spinnaker 3 Microplate Mover.