Search
View additional product information for CTS™ RevitaCell™ Supplement (100X) - FAQs (A4238401)
3 product FAQs found
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems. Learn more here.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
CTS RevitaCell Supplement (100X) should be used in combination with cell culture medium as a post-thaw recovery solution to improve cell viability. It can also be used in combination with CTS Essential 8 Medium (Cat. No. A2656101) when performing single cell passaging procedures.
Find additional tips, troubleshooting help, and resources within our Stem Cell Culture Support Center
The CTS PSC Cryopreservation Kit (Cat. No. A4239301) provides the same efficient recovery, consistency, and reduced variability as the PSC Cryopreservation Kit (Cat. No. A2644601). However, these two kits have different intended use statements, i.e., For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals and For Research Use Only, respectively. Furthermore, the CTS PSC Cryopreservation Kit has full USP sterility testing info and a Drug Master file (DMF), whereas the sterility testing of the PSC Cryopreservation is via a modified USP method, and it does not have a DMF.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.