Search
View additional product information for Calcium Crimson™, AM, cell permeant dye - FAQs (C3018)
3 product FAQs found
The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
This is a known drawback of Calcium Crimson, AM (as well as Calcium Orange, AM and Fura Red, AM, which are also in the same emission range). You can try increasing the concentration and washing out of any unlabeled dye from the media, to try to get better signal-to-background. If that fails, we recommend using Rhod-3 AM instead, which has a much better change in signal in that wavelength.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.