Di-8-ANEPPS - FAQs
View additional product information for Di-8-ANEPPS - FAQs (D3167)
4 product FAQs found
What are the excitation/emission maxima for Di-8-ANEPPS (Cat. No. D3167)?
Di-8-ANEPPS dye (Cat. No. D3167) is essent­ially nonfluorescent until bound to membranes. The excitation/emission maxima for Di-8-ANEPPS when bound to model phospholipid membranes are ~465/635 nm but spectral properties are highly dependent on the environment.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
I am seeing high background outside of my neuronal cells when using membrane potential indicators. What can I do to reduce background?
If you use our FluoVolt Membrane Potential Kit (Cat. No. F10488), the kit provides a background suppressor to reduce this problem. For other indicators, consider the use of BackDrop Background Suppressor (Cat no. R37603, B10511, and B10512).
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
What is the difference between fast and slow-response membrane potential probes?
Molecules that change their structure in response to the surrounding electric field can function as fast-response probes for the detection of transient (millisecond) potential changes. Slow-response dyes function by entering depolarized cells and binding to proteins or membranes. Increased depolarization results in additional dye influx and an increase in fluorescence, while hyperpolarization is indicated by a decrease in fluorescence. Fast-response probes are commonly used to image electrical activity from intact heart tissues or measure membrane potential changes in response to pharmacological stimuli. Slow-responding probes are often used to explore mitochondrial function and cell viability.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
What type of membrane potential indicators do you offer and how should I choose one for my experiment?
A membrane potential indicator selection guide can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-viability-and-regulation/ion-indicators/membrane-potential-indicators.html).
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.