I am labeling brain cryosections with a NeuroTrace Nissl Stain. Is this compatible with antibody labeling?
Yes. We have done this successfully with an anti-GFAP primary and an Alexa Fluor secondary antibody. We would recommend labeling with the primary and secondary antibodies first, then following up with the standard NeuroTrace Nissl Stain protocol.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.