Vybrant™ Phagocytosis Assay Kit - FAQs
View additional product information for Vybrant™ Phagocytosis Assay Kit - FAQs (V6694)
2 product FAQs found
What is phagocytosis?
Phagocytosis is an important mechanism for nourishment in unicellular organisms and for defense against infection in higher vertebrates. The process of phagocytosis can be observed and quantitated in human polynuclear cells and mouse macrophages by following the internalization of a foreign particle such as fluorescently labeled immune complexes and bacterial particles.
Which medium should I use for culturing cells to be used with the Vybrant Phagocytosis Assay Kit (Cat. No. V6694)?
We recommend using any medium that is optimal for culturing your cell(s) of interest. Avoid media that have extremes of pH, heavy metals or strong oxidizing/reducing agents as these may degrade the dye fluorescein on the BioParticles and may degrade or precipitate Trypan blue dye.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.