Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCMH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101390.2 | NP_563966.1 | 1 - 2 | 560 | 105 | |
NM_202105.2 | NP_973834.1 | 2 - 3 | 439 | 105 | ||
GenBank mRNA | AY050392.1 | - | 1 - 2 | 562 | 105 | |
AY085498.1 | - | 2 - 3 | 419 | 105 | ||
AY097355.1 | - | 1 - 2 | 139 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |