Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATNTT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101419.3 | NP_173003.1 | 1 - 2 | 912 | 92 | |
GenBank mRNA | AY081350.1 | - | 1 - 2 | 912 | 92 | |
AY084374.1 | - | 1 - 2 | 888 | 92 | ||
AY128844.1 | - | 1 - 2 | 776 | 92 | ||
X94626.2 | - | 857 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |