Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENP-C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101434.4 | NP_173018.2 | 3 - 4 | 608 | 67 | |
GenBank mRNA | AY693798.1 | - | 3 - 4 | 410 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |