Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084073.1 | NP_001077542.1 | 4 - 5 | 1427 | 78 | |
NM_101437.5 | NP_173021.1 | 4 - 5 | 1491 | 78 | ||
GenBank mRNA | BT002481.1 | - | 4 - 5 | 1427 | 78 | |
M81892.1 | - | 4 - 5 | 1457 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |