Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101438.2 | NP_173022.1 | 1 | 879 | 69 | |
GenBank mRNA | AY065255.1 | - | 1 | 879 | 69 | |
AY117311.1 | - | 1 | 850 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |