Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PnsB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101466.3 | NP_563986.1 | 2 - 3 | 744 | 71 | |
GenBank mRNA | AY057703.1 | - | 2 - 3 | 591 | 71 | |
AY084966.1 | - | 2 - 3 | 591 | 71 | ||
AY124875.1 | - | 2 - 3 | 575 | 71 | ||
KJ139005.1 | - | 2 - 3 | 575 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |