Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPCP-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101496.3 | NP_173080.1 | 11 - 12 | 1212 | 152 | |
GenBank mRNA | BT021096.1 | - | 11 - 12 | 1092 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |