Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATKDSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035978.2 | NP_001031055.1 | 3 - 4 | 235 | 118 | |
NM_001084078.1 | NP_001077547.1 | 3 - 4 | 175 | 118 | ||
NM_101500.4 | NP_173084.1 | 2 - 3 | 424 | 118 | ||
GenBank mRNA | AK175407.1 | - | 3 - 4 | 207 | 118 | |
BT010438.1 | - | 2 - 3 | 132 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |