Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198089.1 | NP_001185018.1 | 10 - 11 | 2120 | 117 | |
NM_101564.4 | NP_173147.2 | 9 - 10 | 2167 | 117 | ||
GenBank mRNA | BT046180.1 | - | 9 - 10 | 1829 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |