Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101587.2 | NP_173169.2 | 3 - 4 | 287 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |