Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AlaAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101591.6 | NP_173173.3 | 12 - 13 | 1746 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |