Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101603.3 | NP_564021.1 | 7 - 8 | 2190 | 75 | |
GenBank mRNA | AY075625.1 | - | 7 - 8 | 2102 | 75 | |
BT006348.1 | - | 7 - 8 | 2001 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |