Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084086.1 | NP_001077555.1 | 11 - 12 | 1176 | 112 | |
NM_101609.4 | NP_173190.1 | 11 - 12 | 1251 | 112 | ||
GenBank mRNA | BT021092.1 | - | 10 - 11 | 1114 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |