Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKD1;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101666.3 | NP_173244.1 | 1 - 2 | 328 | 83 | |
GenBank mRNA | AB047274.1 | - | 1 - 2 | 211 | 83 | |
AY099677.1 | - | 1 - 2 | 300 | 83 | ||
AY128857.1 | - | 1 - 2 | 99 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |