Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1G18070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035986.1 | NP_001031063.1 | 16 - 17 | 1421 | 68 | |
NM_001198097.1 | NP_001185026.1 | 16 - 17 | 1459 | 68 | ||
NM_101669.3 | NP_173247.1 | 16 - 17 | 1470 | 68 | ||
GenBank mRNA | AY120769.1 | - | 16 - 17 | 1405 | 68 | |
BT008889.1 | - | 15 - 16 | 1297 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |