Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMPK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198098.1 | NP_001185027.1 | 6 - 7 | 1154 | 95 | |
NM_101675.4 | NP_173253.1 | 6 - 7 | 1166 | 95 | ||
NM_179354.3 | NP_849685.1 | 5 - 6 | 1246 | 95 | ||
GenBank mRNA | AY045931.1 | - | 6 - 7 | 1076 | 95 | |
AY142618.1 | - | 5 - 6 | 998 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |