Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332355.1 | NP_001321300.1 | 2 - 3 | 1068 | 63 | |
NM_101723.4 | NP_564059.1 | 2 - 3 | 1068 | 63 | ||
GenBank mRNA | AK228238.1 | - | 2 - 3 | 403 | 63 | |
AY087143.1 | - | 2 - 3 | 410 | 63 | ||
BT006195.1 | - | 2 - 3 | 293 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |