Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332357.1 | NP_001321210.1 | 6 - 7 | 1835 | 151 | |
NM_001332358.1 | NP_001321211.1 | 6 - 7 | 1654 | 151 | ||
NM_101725.3 | NP_173302.2 | 6 - 7 | 1835 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |